Product Details

SNP ID
rs144536822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7923504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTGCTCCGCACTCGACACCCC[C/T]AGCAAGACAGAGCTGACACCCTCAC
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
There are no transcripts associated with this gene.

Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004732.3 1123 Silent Mutation CTA,CTG L363L NP_004723.2
XM_011524068.1 1123 Intron XP_011522370.1
XM_017025304.1 1123 Silent Mutation CTA,CTG L343L XP_016880793.1
XM_017025305.1 1123 Intron XP_016880794.1
XM_017025306.1 1123 Silent Mutation CTA,CTG L285L XP_016880795.1
Gene
LOC284023
Gene Name
uncharacterized LOC284023
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
There are no transcripts associated with this gene.

View Full Product Details