Product Details

SNP ID
rs144589585
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51171574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGAAGAACTGGTTGACTACAA[C/G]TCTTGTGCTCATGACTGGGTCTATG
Phenotype
MIM: 156490 MIM: 156491
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
MBTD1 PubMed Links
Additional Information
For this assay, SNP(s) [rs201931229] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MBTD1
Gene Name
mbt domain containing 1
There are no transcripts associated with this gene.

Gene
NME1
Gene Name
NME/NM23 nucleoside diphosphate kinase 1
There are no transcripts associated with this gene.

Gene
NME1-NME2
Gene Name
NME1-NME2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018136.2 546 Missense Mutation AAC,AAG N258K NP_001018146.1
Gene
NME2
Gene Name
NME/NM23 nucleoside diphosphate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018137.2 546 Missense Mutation AAC,AAG N143K NP_001018147.1
NM_001018138.1 546 Intron NP_001018148.1
NM_001018139.2 546 Intron NP_001018149.1
NM_001198682.1 546 UTR 3 NP_001185611.1
NM_002512.3 546 Missense Mutation AAC,AAG N143K NP_002503.1

View Full Product Details