Product Details

SNP ID
rs144606825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41046662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGGTCTTAATAGATGAGCCCA[A/G]TGCTCGGTTCCCTGCTTGGTTGATG
Phenotype
MIM: 608819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP1-1 PubMed Links

Gene Details

Gene
KRTAP1-1
Gene Name
keratin associated protein 1-1
There are no transcripts associated with this gene.

Gene
KRTAP2-1
Gene Name
keratin associated protein 2-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123387.1 Intron NP_001116859.1
Gene
KRTAP2-2
Gene Name
keratin associated protein 2-2
There are no transcripts associated with this gene.

View Full Product Details