Product Details

SNP ID
rs144719287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63820323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGTGCATTGATTTCCCGCCAGC[A/G]TTTCCGGTAATGCTCCACCTCCTTC
Phenotype
MIM: 613369 MIM: 601681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX42 PubMed Links

Gene Details

Gene
DDX42
Gene Name
DEAD-box helicase 42
There are no transcripts associated with this gene.

Gene
FTSJ3
Gene Name
FtsJ homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017647.3 2834 Missense Mutation CGC,TGC R730C NP_060117.3
Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
There are no transcripts associated with this gene.

View Full Product Details