Product Details

SNP ID
rs144782689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41524188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAATCTCAAGACCCTGAAGGGT[A/G]CGCCGCAGGTCAGTAACCTCGGACC
Phenotype
MIM: 148030 MIM: 148020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT15 PubMed Links

Gene Details

Gene
KRT15
Gene Name
keratin 15
There are no transcripts associated with this gene.

Gene
KRT19
Gene Name
keratin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002276.4 Intron NP_002267.2
Gene
MIR6510
Gene Name
microRNA 6510
There are no transcripts associated with this gene.

View Full Product Details