Product Details

SNP ID
rs144789524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41515526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGTAGCGATCTCCTGCTCCAGC[C/T]GTGTCTTTATGTCAAGCAGCATCTT
Phenotype
MIM: 148065 MIM: 148030 MIM: 148020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT13 PubMed Links

Gene Details

Gene
KRT13
Gene Name
keratin 13
There are no transcripts associated with this gene.

Gene
KRT15
Gene Name
keratin 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002275.3 1451 Missense Mutation CAG,CGG Q398R NP_002266.2
XM_011524784.2 1451 Missense Mutation CAG,CGG Q405R XP_011523086.1
XM_017024614.1 1451 Missense Mutation CAG,CGG Q405R XP_016880103.1
Gene
KRT19
Gene Name
keratin 19
There are no transcripts associated with this gene.

Gene
MIR6510
Gene Name
microRNA 6510
There are no transcripts associated with this gene.

View Full Product Details