Product Details

SNP ID
rs144889411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16002721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTTTGAGGGTGAAAGCAAAAGG[A/G]AATTTTAAAGTTTTGAAATAAGTAG
Phenotype
MIM: 613814 MIM: 614535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC19 PubMed Links

Gene Details

Gene
TTC19
Gene Name
tetratricopeptide repeat domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271420.1 Intron NP_001258349.1
NM_017775.3 Intron NP_060245.3
XM_017024801.1 Intron XP_016880290.1
XM_017024802.1 Intron XP_016880291.1
Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
There are no transcripts associated with this gene.

View Full Product Details