Product Details
- SNP ID
-
rs144890975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:215760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTGGCACCCAGAGAGGTGGCCC[A/G]GCGGGTGGGTGAACCCCATCCTGGG
- Phenotype
-
MIM: 604881
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPH3AL
PubMed Links
Gene Details
- Gene
- RPH3AL
- Gene Name
- rabphilin 3A-like (without C2 domains)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190411.1 |
882 |
Missense Mutation |
CCG,CTG |
P257L |
NP_001177340.1 |
NM_001190412.1 |
882 |
Missense Mutation |
CCG,CTG |
P228L |
NP_001177341.1 |
NM_001190413.1 |
882 |
Missense Mutation |
CCG,CTG |
P228L |
NP_001177342.1 |
NM_006987.3 |
882 |
Missense Mutation |
CCG,CTG |
P257L |
NP_008918.1 |
XM_011535364.2 |
882 |
Missense Mutation |
CCG,CTG |
P263L |
XP_011533666.1 |
XM_011535365.2 |
882 |
Missense Mutation |
CCG,CTG |
P263L |
XP_011533667.1 |
XM_011535366.2 |
882 |
Missense Mutation |
CCG,CTG |
P263L |
XP_011533668.1 |
XM_011535367.2 |
882 |
Missense Mutation |
CCG,CTG |
P257L |
XP_011533669.1 |
XM_011535368.2 |
882 |
Missense Mutation |
CCG,CTG |
P228L |
XP_011533670.1 |
XM_011535369.2 |
882 |
Missense Mutation |
CCG,CTG |
P181L |
XP_011533671.1 |
XM_017025367.1 |
882 |
Intron |
|
|
XP_016880856.1 |
XM_017025368.1 |
882 |
Intron |
|
|
XP_016880857.1 |
XM_017025369.1 |
882 |
Missense Mutation |
CCG,CTG |
P181L |
XP_016880858.1 |
XM_017025370.1 |
882 |
Missense Mutation |
CCG,CTG |
P175L |
XP_016880859.1 |
XM_017025371.1 |
882 |
Intron |
|
|
XP_016880860.1 |
View Full Product Details