Product Details

SNP ID
rs145028500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15708172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAATAGGGCTTCCAGTTTCCAAA[C/T]CTGAGAGCTACAACTTGGAGAATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF286A PubMed Links

Gene Details

Gene
ZNF286A
Gene Name
zinc finger protein 286A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130842.1 Intron NP_001124314.1
NM_001288642.1 Intron NP_001275571.1
NM_001288643.1 Intron NP_001275572.1
NM_001288644.1 Intron NP_001275573.1
NM_001288645.1 Intron NP_001275574.1
NM_001288646.1 Intron NP_001275575.1
NM_001288647.1 Intron NP_001275576.1
NM_001288648.1 Intron NP_001275577.1
NM_001288649.1 Intron NP_001275578.1
NM_020652.2 Intron NP_065703.1

View Full Product Details