Product Details

SNP ID
rs145109964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81845187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGCCACCGCTCTCCAGGAAT[G/T]TCTTAAAACCATCCAGCGTGCGTTC
Phenotype
MIM: 176790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
P4HB PubMed Links

Gene Details

Gene
P4HB
Gene Name
prolyl 4-hydroxylase subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000918.3 1600 Missense Mutation AAA,ACA K468T NP_000909.2

View Full Product Details