Product Details

SNP ID
rs145177851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39212390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCCTTGCTTCTGCCCACGCCCA[C/T]GCAGATCTGAGCCAGAGAGACATGG
Phenotype
MIM: 180466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL19 PubMed Links

Gene Details

Gene
RPL19
Gene Name
ribosomal protein L19
There are no transcripts associated with this gene.

Gene
STAC2
Gene Name
SH3 and cysteine rich domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198993.3 1273 Missense Mutation ATG,GTG M380V NP_945344.1
XM_017024580.1 1273 Missense Mutation ATG,GTG M385V XP_016880069.1
XM_017024581.1 1273 Missense Mutation ATG,GTG M330V XP_016880070.1
XM_017024582.1 1273 Intron XP_016880071.1
XM_017024583.1 1273 Intron XP_016880072.1

View Full Product Details