Product Details

SNP ID
rs145247799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61953049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGAAGCAGGAAGGATATTAAT[A/G]ATATCAGGGTCAAGATCATCTCCTG
Phenotype
MIM: 603808
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED13 PubMed Links

Gene Details

Gene
MED13
Gene Name
mediator complex subunit 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005121.2 5525 Silent Mutation ATC,ATT I2011I NP_005112.2
XM_011525551.2 5525 Silent Mutation ATC,ATT I1958I XP_011523853.1
XM_011525552.2 5525 Intron XP_011523854.1
XM_011525553.2 5525 Silent Mutation ATC,ATT I1788I XP_011523855.1

View Full Product Details