Product Details

SNP ID
rs145349423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81806245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCCGCAGGGAACGGAGGACGCT[A/C]ACACTTCTGCACCTCCTGCCTCACT
Phenotype
MIM: 138033
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000160.4 Intron NP_000151.1
XM_006722277.1 Intron XP_006722340.1
XM_011523539.1 Intron XP_011521841.1
XM_017024446.1 Intron XP_016879935.1
XM_017024447.1 Intron XP_016879936.1

View Full Product Details