Product Details

SNP ID
rs145372121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36599049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCCCAGACCCACATCCGAGCCA[C/T]GGTGAGGCTGTGGCCTAGCCCTGGT
Phenotype
MIM: 616159 MIM: 612275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS11 PubMed Links

Gene Details

Gene
DHRS11
Gene Name
dehydrogenase/reductase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024308.3 795 Missense Mutation ACG,ATG T194M NP_077284.2
XM_005257658.2 795 Missense Mutation ACG,ATG T194M XP_005257715.1
XM_011525233.2 795 Missense Mutation ACG,ATG T124M XP_011523535.1
Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
There are no transcripts associated with this gene.

Gene
MRM1
Gene Name
mitochondrial rRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details