Product Details

SNP ID
rs145559227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29629658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAAGACAGTGACAATATTATGT[A/C]CATAAATACTGTGTACAAACTTTAG
Phenotype
MIM: 606779
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CORO6 PubMed Links

Gene Details

Gene
CORO6
Gene Name
coronin 6
There are no transcripts associated with this gene.

Gene
SSH2
Gene Name
slingshot protein phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282129.1 6255 UTR 3 NP_001269058.1
NM_001282130.1 6255 Intron NP_001269059.1
NM_001282131.1 6255 Intron NP_001269060.1
NM_033389.3 6255 UTR 3 NP_203747.2
XM_005258058.3 6255 UTR 3 XP_005258115.1
XM_005258059.2 6255 UTR 3 XP_005258116.1
XM_005258060.3 6255 UTR 3 XP_005258117.1
XM_006722149.3 6255 UTR 3 XP_006722212.1
XM_011525402.2 6255 UTR 3 XP_011523704.1
XM_011525404.2 6255 UTR 3 XP_011523706.1
XM_011525405.2 6255 UTR 3 XP_011523707.1
XM_011525406.2 6255 UTR 3 XP_011523708.1
XM_011525407.1 6255 UTR 3 XP_011523709.1
XM_017025251.1 6255 UTR 3 XP_016880740.1
XM_017025252.1 6255 UTR 3 XP_016880741.1
XM_017025253.1 6255 Intron XP_016880742.1
XM_017025254.1 6255 Intron XP_016880743.1
XM_017025255.1 6255 Intron XP_016880744.1

View Full Product Details