Product Details

SNP ID
rs145584568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59629404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTAGTGAGAGACAAATTTTACA[C/T]ATCTAAAATTCTCCTGTGTATGGCC
Phenotype
MIM: 118955
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLTC PubMed Links

Gene Details

Gene
CLTC
Gene Name
clathrin heavy chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288653.1 Intron NP_001275582.1
NM_004859.3 Intron NP_004850.1
XM_005257012.3 Intron XP_005257069.1

View Full Product Details