Product Details

SNP ID
rs145752810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1496334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGACGTACCTGGTTCAGGTTGTC[A/G]CTGCAGGAGACCTTGCTGTCCCCGA
Phenotype
MIM: 607875 MIM: 606538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INPP5K PubMed Links

Gene Details

Gene
INPP5K
Gene Name
inositol polyphosphate-5-phosphatase K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135642.1 1339 Silent Mutation AGC,AGT S314S NP_001129114.1
NM_016532.3 1339 Silent Mutation AGC,AGT S390S NP_057616.2
NM_130766.2 1339 Silent Mutation AGC,AGT S314S NP_570122.1
XM_005256683.2 1339 Silent Mutation AGC,AGT S314S XP_005256740.1
XM_005256685.1 1339 Silent Mutation AGC,AGT S298S XP_005256742.1
XM_005256686.1 1339 Silent Mutation AGC,AGT S298S XP_005256743.1
XM_011523934.1 1339 Silent Mutation AGC,AGT S314S XP_011522236.1
XM_011523936.2 1339 Silent Mutation AGC,AGT S255S XP_011522238.1
XM_017024755.1 1339 Silent Mutation AGC,AGT S314S XP_016880244.1
XM_017024756.1 1339 Silent Mutation AGC,AGT S314S XP_016880245.1
XM_017024757.1 1339 Silent Mutation AGC,AGT S298S XP_016880246.1
XM_017024758.1 1339 Silent Mutation AGC,AGT S255S XP_016880247.1
XM_017024759.1 1339 Silent Mutation AGC,AGT S255S XP_016880248.1
Gene
MYO1C
Gene Name
myosin IC
There are no transcripts associated with this gene.

View Full Product Details