Product Details

SNP ID
rs145760222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7688835 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATCCTGTGGCTGGCTCAGCT[A/G]TGTCCCAGGAGCTACGGGAGGGGGA
Phenotype
MIM: 191170 MIM: 612661
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TP53 PubMed Links

Gene Details

Gene
TP53
Gene Name
tumor protein p53
There are no transcripts associated with this gene.

Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143990.1 342 Missense Mutation ATG,GTG M63V NP_001137462.1
NM_001143991.1 342 Missense Mutation ATG,GTG M63V NP_001137463.1
NM_001143992.1 342 Missense Mutation ATG,GTG M63V NP_001137464.1
NM_018081.2 342 Missense Mutation ATG,GTG M63V NP_060551.2
XM_011523952.2 342 UTR 5 XP_011522254.1
XM_017024812.1 342 Missense Mutation ATG,GTG M63V XP_016880301.1

View Full Product Details