Product Details

SNP ID
rs145849983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36494001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTTCTTTGCAGAATTTAAAGA[A/G]TTTAGGTAAGTCTGTGCTATGCTTG
Phenotype
MIM: 604500
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO19 PubMed Links

Gene Details

Gene
MYO19
Gene Name
myosin XIX
There are no transcripts associated with this gene.

Gene
ZNHIT3
Gene Name
zinc finger HIT-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281432.1 354 Missense Mutation AAT,AGT N94S NP_001268361.1
NM_001281433.1 354 UTR 3 NP_001268362.1
NM_001281434.1 354 Intron NP_001268363.1
NM_004773.3 354 Missense Mutation AAT,AGT N94S NP_004764.1

View Full Product Details