Product Details

SNP ID
rs145850889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7342064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGTCCACCAGAGCCCATGAT[A/G]GCGGTATGCGGAGGGTCTGCATCTG
Phenotype
MIM: 607763 MIM: 609848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAP1 PubMed Links

Gene Details

Gene
ACAP1
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014716.3 434 Missense Mutation ATA,ATG I76M NP_055531.1
Gene
KCTD11
Gene Name
potassium channel tetramerization domain containing 11
There are no transcripts associated with this gene.

View Full Product Details