Product Details

SNP ID
rs145861447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40750525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGCTCCTCCAGGGCCCCGATCT[A/G]AGCCTGGATCTGCGCCAGCTGCGCA
Phenotype
MIM: 616646
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT25 PubMed Links

Gene Details

Gene
KRT25
Gene Name
keratin 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181534.3 1079 Nonsense Mutation CAG,TAG Q344* NP_853512.1
XM_011524414.1 1079 Nonsense Mutation CAG,TAG Q342* XP_011522716.1

View Full Product Details