Product Details

SNP ID
rs145873506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51168275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACACCTGAAGCAGCACTACATT[A/G]ACCTGAAAGACCGACCATTCTTCCC
Phenotype
MIM: 156490 MIM: 156491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBTD1 PubMed Links

Gene Details

Gene
MBTD1
Gene Name
mbt domain containing 1
There are no transcripts associated with this gene.

Gene
NME1
Gene Name
NME/NM23 nucleoside diphosphate kinase 1
There are no transcripts associated with this gene.

Gene
NME1-NME2
Gene Name
NME1-NME2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018136.2 390 Missense Mutation AAC,GAC N169D NP_001018146.1
Gene
NME2
Gene Name
NME/NM23 nucleoside diphosphate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018137.2 390 Missense Mutation AAC,GAC N54D NP_001018147.1
NM_001018138.1 390 Intron NP_001018148.1
NM_001018139.2 390 Intron NP_001018149.1
NM_001198682.1 390 Missense Mutation AAC,GAC N54D NP_001185611.1
NM_002512.3 390 Missense Mutation AAC,GAC N54D NP_002503.1

View Full Product Details