Product Details

SNP ID
rs145991218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29575138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGATCTTCTCTGAGCAGGGCA[C/T]GAAGCTGCGGGGAGAAGGGAGGCTA
Phenotype
MIM: 608434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABHD15 PubMed Links

Gene Details

Gene
ABHD15
Gene Name
abhydrolase domain containing 15
There are no transcripts associated with this gene.

Gene
GIT1
Gene Name
GIT ArfGAP 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085454.1 2228 Missense Mutation ATG,GTG M681V NP_001078923.1
NM_014030.3 2228 Missense Mutation ATG,GTG M672V NP_054749.2
XM_011524684.1 2228 Missense Mutation ATG,GTG M681V XP_011522986.1
XM_011524685.1 2228 Missense Mutation ATG,GTG M672V XP_011522987.1
Gene
TP53I13
Gene Name
tumor protein p53 inducible protein 13
There are no transcripts associated with this gene.

View Full Product Details