Product Details

SNP ID
rs146139123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41853777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGCTTGGCACTCGCCTCATG[C/T]TCAGGGCGTGACGATGTATCTCTTG
Phenotype
MIM: 608778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL10 PubMed Links

Gene Details

Gene
KLHL10
Gene Name
kelch like family member 10
There are no transcripts associated with this gene.

Gene
KLHL11
Gene Name
kelch like family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018143.2 2151 Missense Mutation AAC,AGC N697S NP_060613.1

View Full Product Details