Product Details

SNP ID
rs146175295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40656510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTGCTTAGTCTCTAAACACCC[C/T]TCATCAGTGGCTGCTAAATGAAGAT
Phenotype
MIM: 603111
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT222 PubMed Links

Gene Details

Gene
KRT222
Gene Name
keratin 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152349.2 845 Silent Mutation GAA,GAG E260E NP_689562.1
Gene
SMARCE1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1
There are no transcripts associated with this gene.

View Full Product Details