Product Details

SNP ID
rs146218228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47809228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAACTCCAGTACTTGGCCTGGGGT[C/G]GGGAAGGCAGGGTTTAGGGAGGTGT
Phenotype
MIM: 606735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OSBPL7 PubMed Links

Gene Details

Gene
OSBPL7
Gene Name
oxysterol binding protein like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145798.2 Intron NP_665741.1
XM_017024122.1 Intron XP_016879611.1
XM_017024123.1 Intron XP_016879612.1
XM_017024124.1 Intron XP_016879613.1
XM_017024125.1 Intron XP_016879614.1

View Full Product Details