Product Details

SNP ID
rs146268308
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:13496758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGCGCGGGGGCCTCCCGCGTGAC[A/G]AAGTAACTGGGCGTCTTCTCCATGG
Phenotype
MIM: 604057
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HS3ST3A1 PubMed Links
Additional Information
For this assay, SNP(s) [rs8080565] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HS3ST3A1
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 3A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006042.2 1473 Silent Mutation TTC,TTT F220F NP_006033.1
XM_011524114.2 1473 Silent Mutation TTC,TTT F21F XP_011522416.1
XM_017025480.1 1473 Intron XP_016880969.1

View Full Product Details