Product Details

SNP ID
rs146291979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61591180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTTTCTGTGGCTTCGCCCGGCA[G/T]TTTGTGCAGGGAACGCGGAAGCAAG
Phenotype
MIM: 609274
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NACA2 PubMed Links

Gene Details

Gene
NACA2
Gene Name
nascent polypeptide associated complex alpha subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199290.3 Intron NP_954984.1

View Full Product Details