Product Details

SNP ID
rs146314563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5179831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGGTGCCAGTCAGAAGCATGA[C/T]TTGTGTTGTACATGCCTTTATGTAG
Phenotype
MIM: 604334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP6 PubMed Links

Gene Details

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
There are no transcripts associated with this gene.

Gene
ZNF594
Gene Name
zinc finger protein 594
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032530.1 4895 UTR 3 NP_115919.1
XM_005256827.2 4895 UTR 3 XP_005256884.1

View Full Product Details