Product Details

SNP ID
rs146555797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1423517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAAACTGTTGAACTATACTCAG[C/T]TGAAGTCCTCATCGGGATTCTGTTG
Phenotype
MIM: 164762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRK PubMed Links

Gene Details

Gene
CRK
Gene Name
CRK proto-oncogene, adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005206.4 1061 UTR 3 NP_005197.3
NM_016823.3 1061 Missense Mutation AAC,AGC N304S NP_058431.2

View Full Product Details