Product Details

SNP ID
rs146596349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42405001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGCGACGTCTTCAGTTTCTCGC[C/G]CCGCTCGGCGGGCACCAGGCGCGTG
Phenotype
MIM: 603198
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PTRF PubMed Links

Gene Details

Gene
PTRF
Gene Name
polymerase I and transcript release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012232.5 1082 Missense Mutation CGC,GGC R287G NP_036364.2
XM_005257242.3 1082 Intron XP_005257299.1

View Full Product Details