Product Details

SNP ID
rs146642281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50383034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACCCACTGGAGGATCTCATGG[C/T]GGCGTAGACCCTGGACCGCATTGTC
Phenotype
MIM: 610885 MIM: 614854 MIM: 611837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EME1 PubMed Links

Gene Details

Gene
EME1
Gene Name
essential meiotic structure-specific endonuclease 1
There are no transcripts associated with this gene.

Gene
LRRC59
Gene Name
leucine rich repeat containing 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018509.3 1114 Missense Mutation CAC,CGC H293R NP_060979.2
Gene
MRPL27
Gene Name
mitochondrial ribosomal protein L27
There are no transcripts associated with this gene.

View Full Product Details