Product Details

SNP ID
rs146681986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42985710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGCTGTAGACTTCTAGTTTAAGA[C/T]GCAGCCTTGCCCCTACAAATGATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGES3L PubMed Links

Gene Details

Gene
PTGES3L
Gene Name
prostaglandin E synthase 3 like
There are no transcripts associated with this gene.

Gene
PTGES3L-AARSD1
Gene Name
PTGES3L-AARSD1 readthrough
There are no transcripts associated with this gene.

Gene
RUNDC1
Gene Name
RUN domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321381.1 Intron NP_001308310.1
NM_173079.3 Intron NP_775102.2
XM_005257078.3 Intron XP_005257135.1
XM_005257080.3 Intron XP_005257137.1

View Full Product Details