Product Details

SNP ID
rs146797327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3610102 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAACTTTTCCAGAAGTCCATA[C/T]GACCCGAGTGTGGGCTTCAGGTCTC
Phenotype
MIM: 605060 MIM: 602076
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHPK PubMed Links

Gene Details

Gene
SHPK
Gene Name
sedoheptulokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013276.2 1998 UTR 3 NP_037408.2
Gene
TRPV1
Gene Name
transient receptor potential cation channel subfamily V member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018727.5 1998 Intron NP_061197.4
NM_080704.3 1998 Intron NP_542435.2
NM_080705.3 1998 Intron NP_542436.2
NM_080706.3 1998 Intron NP_542437.2

View Full Product Details