Product Details

SNP ID
rs147051753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42670581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGGAGAGCCTCACCTGCTCAA[A/G]GAGGAAAGGCAGTTCGTGACCGTCT
Phenotype
MIM: 600240 MIM: 602346 MIM: 605785
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCR10 PubMed Links

Gene Details

Gene
CCR10
Gene Name
C-C motif chemokine receptor 10
There are no transcripts associated with this gene.

Gene
CNTNAP1
Gene Name
contactin associated protein 1
There are no transcripts associated with this gene.

Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024927.4 1849 Missense Mutation CTT,TTT L516F NP_079203.3
XM_017025113.1 1849 Missense Mutation CTT,TTT L605F XP_016880602.1
XM_017025114.1 1849 Missense Mutation CTT,TTT L602F XP_016880603.1
XM_017025115.1 1849 Missense Mutation CTT,TTT L561F XP_016880604.1
XM_017025116.1 1849 Missense Mutation CTT,TTT L560F XP_016880605.1
XM_017025117.1 1849 Missense Mutation CTT,TTT L545F XP_016880606.1
XM_017025118.1 1849 Missense Mutation CTT,TTT L513F XP_016880607.1
Gene
TUBG2
Gene Name
tubulin gamma 2
There are no transcripts associated with this gene.

View Full Product Details