Product Details

SNP ID
rs147085726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59859502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTACTTTTGTACTTAAGCATAA[C/G]CATTTATTACCAAAATCCTCATATT
Phenotype
MIM: 607344
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TUBD1 PubMed Links

Gene Details

Gene
TUBD1
Gene Name
tubulin delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193609.1 2470 UTR 3 NP_001180538.1
NM_001193610.1 2470 UTR 3 NP_001180539.1
NM_001193611.1 2470 UTR 3 NP_001180540.1
NM_001193612.1 2470 UTR 3 NP_001180541.1
NM_001193613.1 2470 UTR 3 NP_001180542.1
NM_016261.3 2470 UTR 3 NP_057345.2
XM_005257425.1 2470 UTR 3 XP_005257482.1
XM_011524871.2 2470 Intron XP_011523173.1
XM_017024716.1 2470 UTR 3 XP_016880205.1
XM_017024717.1 2470 UTR 3 XP_016880206.1
XM_017024718.1 2470 UTR 3 XP_016880207.1
XM_017024719.1 2470 UTR 3 XP_016880208.1
XM_017024720.1 2470 UTR 3 XP_016880209.1

View Full Product Details