Product Details

SNP ID
rs147160209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82460037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGATGACCAAGAGAATGTGTCA[A/G]CCGATGCACCGAGTCCAGCCCAGGA
Phenotype
MIM: 605349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf62 PubMed Links

Gene Details

Gene
C17orf62
Gene Name
chromosome 17 open reading frame 62
There are no transcripts associated with this gene.

Gene
NARF
Gene Name
nuclear prelamin A recognition factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038618.2 119 UTR 5 NP_001033707.1
NM_001083608.1 119 Missense Mutation ACC,GCC T25A NP_001077077.1
NM_012336.3 119 Missense Mutation ACC,GCC T25A NP_036468.1
NM_031968.2 119 Missense Mutation ACC,GCC T25A NP_114174.1
XM_005256340.3 119 UTR 5 XP_005256397.1
XM_006722278.3 119 Missense Mutation ACC,GCC T25A XP_006722341.1
XM_011523541.2 119 UTR 5 XP_011521843.1
XM_011523543.2 119 UTR 5 XP_011521845.1
XM_011523544.2 119 Missense Mutation ACC,GCC T25A XP_011521846.1
XM_017024449.1 119 UTR 5 XP_016879938.1
XM_017024450.1 119 UTR 5 XP_016879939.1

View Full Product Details