Product Details

SNP ID
rs147171402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5183253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGATGTTTAAGAAAAGCTGTG[C/T]GCCCACTGAAGGTCTTCCCACATCT
Phenotype
MIM: 604334
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100130950 PubMed Links

Gene Details

Gene
LOC100130950
Gene Name
uncharacterized LOC100130950
There are no transcripts associated with this gene.

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
There are no transcripts associated with this gene.

Gene
ZNF594
Gene Name
zinc finger protein 594
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032530.1 1473 Missense Mutation CAC,CGC H335R NP_115919.1
XM_005256827.2 1473 Missense Mutation CAC,CGC H335R XP_005256884.1

View Full Product Details