Product Details

SNP ID
rs147264255
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8880974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCAAAGGGGCAGCTGTTGGAG[C/G]CGATGATCTGCACCTTGTCCACTTT
Phenotype
MIM: 611317
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PIK3R5 PubMed Links

Gene Details

Gene
PIK3R5
Gene Name
phosphoinositide-3-kinase regulatory subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142633.2 2381 Missense Mutation GCC,GGC A809G NP_001136105.1
NM_001251851.1 2381 Missense Mutation GCC,GGC A423G NP_001238780.1
NM_001251852.1 2381 Missense Mutation GCC,GGC A423G NP_001238781.1
NM_001251853.1 2381 Missense Mutation GCC,GGC A423G NP_001238782.1
NM_001251855.1 2381 Missense Mutation GCC,GGC A423G NP_001238784.1
NM_014308.3 2381 Missense Mutation GCC,GGC A809G NP_055123.2
XM_005256580.3 2381 Missense Mutation GCC,GGC A808G XP_005256637.1

View Full Product Details