Product Details

SNP ID
rs147330096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2042912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTTCAAGGAATCCATCCTGCA[A/G]AGGCCCTTGTCATTGCCTTCGCTCC
Phenotype
MIM: 603527 MIM: 610016 MIM: 613487 MIM: 607896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPH1 PubMed Links

Gene Details

Gene
DPH1
Gene Name
diphthamide biosynthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001383.3 1676 UTR 3 NP_001374.3
Gene
MIR132
Gene Name
microRNA 132
There are no transcripts associated with this gene.

Gene
MIR212
Gene Name
microRNA 212
There are no transcripts associated with this gene.

Gene
OVCA2
Gene Name
ovarian tumor suppressor candidate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080822.2 1676 Silent Mutation CAA,CAG Q164Q NP_543012.1

View Full Product Details