Product Details

SNP ID
rs147367790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44320061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGAAGCTTTTGCCGAACTCATA[C/G]GTGCTGATCATGATGGCACAGGAGG
Phenotype
MIM: 605448 MIM: 610820
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RUNDC3A PubMed Links

Gene Details

Gene
RUNDC3A
Gene Name
RUN domain containing 3A
There are no transcripts associated with this gene.

Gene
SLC25A39
Gene Name
solute carrier family 25 member 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143780.2 1110 Silent Mutation ACC,ACG T340T NP_001137252.1
NM_001321240.1 1110 Silent Mutation ACC,ACG T317T NP_001308169.1
NM_001321241.1 1110 Silent Mutation ACC,ACG T338T NP_001308170.1
NM_016016.3 1110 Silent Mutation ACC,ACG T332T NP_057100.1
XM_011524880.2 1110 Silent Mutation ACC,ACG T346T XP_011523182.1
XM_011524882.2 1110 Silent Mutation ACC,ACG T323T XP_011523184.1
XM_011524883.2 1110 Silent Mutation ACC,ACG T245T XP_011523185.1
XM_011524884.1 1110 Silent Mutation ACC,ACG T245T XP_011523186.1
XM_017024737.1 1110 Silent Mutation ACC,ACG T239T XP_016880226.1

View Full Product Details