Product Details

SNP ID
rs147371024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34983100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGAAAACATCACTGGCGTGATG[G/T]AAGACAATTCAGCCAGTGGTCAGAA
Phenotype
MIM: 610730 MIM: 600940
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
There are no transcripts associated with this gene.

Gene
LIG3
Gene Name
DNA ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002311.4 224 Missense Mutation GGA,GTA G32V NP_002302.2
NM_013975.3 224 Missense Mutation GGA,GTA G32V NP_039269.2
XM_005257970.3 224 Missense Mutation GGA,GTA G41V XP_005258027.1
XM_006721896.3 224 Missense Mutation GGA,GTA G41V XP_006721959.1
XM_017024624.1 224 Missense Mutation GGA,GTA G32V XP_016880113.1

View Full Product Details