Product Details

SNP ID
rs147384057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42811163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACACACACCCAAAGAGAAGAGAAG[C/T]CTGGAGTAGGGGTGAGTGAGGGTTA
Phenotype
MIM: 604378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BECN1 PubMed Links

Gene Details

Gene
BECN1
Gene Name
beclin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313998.1 2853 Intron NP_001300927.1
NM_001313999.1 2853 Intron NP_001300928.1
NM_001314000.1 2853 Intron NP_001300929.1
NM_003766.4 2853 Intron NP_003757.1
XM_005257759.2 2853 Intron XP_005257816.1
XM_005257760.3 2853 Intron XP_005257817.1
XM_011525421.2 2853 Intron XP_011523723.1
XM_017025262.1 2853 Intron XP_016880751.1
XM_017025263.1 2853 Intron XP_016880752.1
XM_017025264.1 2853 Intron XP_016880753.1
Gene
CNTD1
Gene Name
cyclin N-terminal domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173478.2 2853 UTR 3 NP_775749.2
XM_005257043.4 2853 Intron XP_005257100.1
XM_005257044.4 2853 Intron XP_005257101.1
XM_005257045.4 2853 Intron XP_005257102.1
XM_006721690.2 2853 Intron XP_006721753.1
XM_011524310.2 2853 Intron XP_011522612.1
XM_011524311.2 2853 Intron XP_011522613.1
XM_017024171.1 2853 Intron XP_016879660.1

View Full Product Details