Product Details

SNP ID
rs147477038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41420920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCATGGCCAGAGGGAGACCCACC[A/G]GTGACAGGGCCACAGCTTGGACAAG
Phenotype
MIM: 604541
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT37 PubMed Links

Gene Details

Gene
KRT37
Gene Name
keratin 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003770.4 1355 Silent Mutation NP_003761.3
Gene
LOC100505782
Gene Name
uncharacterized LOC100505782
There are no transcripts associated with this gene.

View Full Product Details