Product Details

SNP ID
rs147503881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42207631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTTTTGGCCGATCAGGAAACAC[A/G]TAGATAAGGTAATTCAAGTCTCCCA
Phenotype
MIM: 604260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STAT5B PubMed Links

Gene Details

Gene
STAT5B
Gene Name
signal transducer and activator of transcription 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012448.3 1992 Silent Mutation TAC,TAT Y668Y NP_036580.2
XM_005257626.4 1992 Intron XP_005257683.1
XM_017024977.1 1992 Silent Mutation TAC,TAT Y574Y XP_016880466.1

View Full Product Details