Product Details

SNP ID
rs147520731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81811902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCACAGGTGCCCCCATGCTGTT[C/T]GTCGTCCCCTGGGCAGTGGTCAAGT
Phenotype
MIM: 138033
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCGR PubMed Links

Gene Details

Gene
GCGR
Gene Name
glucagon receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000160.4 1129 Silent Mutation TTC,TTT F278F NP_000151.1
XM_006722277.1 1129 Silent Mutation TTC,TTT F278F XP_006722340.1
XM_011523539.1 1129 Silent Mutation TTC,TTT F245F XP_011521841.1
XM_017024446.1 1129 Silent Mutation TTC,TTT F276F XP_016879935.1
XM_017024447.1 1129 Silent Mutation TTC,TTT F156F XP_016879936.1

View Full Product Details