Product Details

SNP ID
rs147629920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7036438 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGGGATGGGTGGTGGTGCTCT[C/T]AGCGTTCTTCCAGTCGGCGCTTGTG
Phenotype
MIM: 608992 MIM: 615765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL6B PubMed Links

Gene Details

Gene
BCL6B
Gene Name
B-cell CLL/lymphoma 6B
There are no transcripts associated with this gene.

Gene
SLC16A11
Gene Name
solute carrier family 16 member 11
There are no transcripts associated with this gene.

Gene
SLC16A13
Gene Name
solute carrier family 16 member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201566.2 364 Missense Mutation TCA,TTA S19L NP_963860.1

View Full Product Details