Product Details
- SNP ID
-
rs147636121
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17477161 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCTCAGCCGCCGCCGGTCCCTGC[A/G]CCTCAACCGCAGCAGTCGCCGGCGC
- Phenotype
-
MIM: 609878
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MED9
PubMed Links
Gene Details
- Gene
- MED9
- Gene Name
- mediator complex subunit 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018019.2 |
176 |
Silent Mutation |
GCA,GCG |
A40A |
NP_060489.1 |
View Full Product Details