Product Details

SNP ID
rs147680117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74358139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCTAATCAATCACCTAGGAAAG[A/G]GGGAGTGGCACACACCCCTCAGGGT
Phenotype
MIM: 609046
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTBD17 PubMed Links

Gene Details

Gene
BTBD17
Gene Name
BTB domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080466.1 Intron NP_001073935.1
XM_011524791.2 Intron XP_011523093.1
XM_017024622.1 Intron XP_016880111.1
Gene
GPR142
Gene Name
G protein-coupled receptor 142
There are no transcripts associated with this gene.

Gene
KIF19
Gene Name
kinesin family member 19
There are no transcripts associated with this gene.

View Full Product Details