Product Details

SNP ID
rs147741483
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41190523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAATGAGTCCAGCTATTGTCTG[A/C]CTTGCTGCCGTCCCACCTGCTGCCA
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
KRTAP4-1 PubMed Links

Gene Details

Gene
KRTAP4-1
Gene Name
keratin associated protein 4-1
There are no transcripts associated with this gene.

Gene
KRTAP9-1
Gene Name
keratin associated protein 9-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190460.1 451 Missense Mutation ACT,CCT T213P NP_001177389.1
XM_005257633.1 451 Missense Mutation ACT,CCT T151P XP_005257690.1

View Full Product Details